Product Details

SNP ID
rs188065910
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:160679029 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAGATCTTCTGGCCTTGAAGTTT[C/T]GCTTGAGTTAAAAGAGCTCATCTCA
Phenotype
MIM: 109530
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD48 PubMed Links

Gene Details

Gene
CD48
Gene Name
CD48 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256030.1 911 Intron NP_001242959.1
NM_001778.3 911 UTR 3 NP_001769.2
XM_005245625.1 911 UTR 3 XP_005245682.1
XM_011510171.1 911 UTR 3 XP_011508473.1
XM_017002867.1 911 UTR 3 XP_016858356.1

View Full Product Details