Product Details

SNP ID
hCV25599998
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:7640879 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGGGGGCTGCTCTGGCCTGATGC[A/C]CCACTCCTGCCTCACCCCAGCCTGC
Phenotype
MIM: 614770 MIM: 601717
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PCP2 PubMed Links

Gene Details

Gene
PCP2
Gene Name
Purkinje cell protein 2
There are no transcripts associated with this gene.

Gene
PET100
Gene Name
PET100 homolog
There are no transcripts associated with this gene.

Gene
STXBP2
Gene Name
syntaxin binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127396.2 Intron NP_001120868.1
NM_001272034.1 Intron NP_001258963.1
NM_006949.3 Intron NP_008880.2

View Full Product Details