Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_005468.2 | 2125 | Silent Mutation | GCC,GCT | A703A | NP_005459.2 |
XM_011544706.2 | 2125 | Silent Mutation | GCC,GCT | A700A | XP_011543008.1 |
XM_011544707.2 | 2125 | Silent Mutation | GCC,GCT | A655A | XP_011543009.1 |
XM_011544708.2 | 2125 | Silent Mutation | GCC,GCT | A650A | XP_011543010.1 |
XM_011544709.2 | 2125 | Intron | XP_011543011.1 | ||
XM_011544710.2 | 2125 | Intron | XP_011543012.1 | ||
XM_011544711.2 | 2125 | Intron | XP_011543013.1 | ||
XM_011544712.2 | 2125 | Intron | XP_011543014.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_013299.3 | 2125 | Intron | NP_037431.3 |