Product Details

SNP ID
rs12031285
Assay Type
Functionally tested
NCBI dbSNP Submissions
44
Location
Chr.1:211663447 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGGCTAGCGCATGCCCAGGATCTG[C/T]CTGCTTTTCAGTTGGTAATTTTTCT
Phenotype
MIM: 604043
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LOC105748977 PubMed Links

Gene Details

Gene
LOC105748977
Gene Name
uncharacterized LOC105748977
There are no transcripts associated with this gene.

Gene
NEK2
Gene Name
NIMA related kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204182.1 1441 Intron NP_001191111.1
NM_001204183.1 1441 Intron NP_001191112.1
NM_002497.3 1441 Silent Mutation AGA,AGG R439R NP_002488.1
XM_005273147.1 1441 Silent Mutation AGA,AGG R431R XP_005273204.1

View Full Product Details