Product Details

SNP ID
rs35271800
Assay Type
Functionally tested
NCBI dbSNP Submissions
21
Location
Chr.1:2055515 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGAGGAAGTGAGAGACATGTGTC[A/G]TCTGCACCAGCAGCACCCGCTCACC
Phenotype
MIM: 176982
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105378591 PubMed Links

Gene Details

Gene
LOC105378591
Gene Name
uncharacterized LOC105378591
There are no transcripts associated with this gene.

Gene
PRKCZ
Gene Name
protein kinase C zeta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033581.1 303 Intron NP_001028753.1
NM_001033582.1 303 Intron NP_001028754.1
NM_001242874.1 303 Intron NP_001229803.1
NM_002744.4 303 Missense Mutation CAT,CGT H49R NP_002735.3
XM_011541773.1 303 Missense Mutation CAT,CGT H49R XP_011540075.1
XM_011541774.1 303 Intron XP_011540076.1
XM_011541775.2 303 Missense Mutation CAT,CGT H49R XP_011540077.1
XM_011541776.1 303 Intron XP_011540078.1
XM_011541778.1 303 Intron XP_011540080.1
XM_017001789.1 303 Missense Mutation CAT,CGT H49R XP_016857278.1
XM_017001790.1 303 Missense Mutation CAT,CGT H49R XP_016857279.1
XM_017001791.1 303 Intron XP_016857280.1
XM_017001792.1 303 Intron XP_016857281.1
XM_017001793.1 303 Intron XP_016857282.1
XM_017001794.1 303 Intron XP_016857283.1
XM_017001795.1 303 Intron XP_016857284.1
XM_017001796.1 303 Intron XP_016857285.1
XM_017001797.1 303 Intron XP_016857286.1
XM_017001798.1 303 Intron XP_016857287.1
XM_017001799.1 303 Intron XP_016857288.1
XM_017001800.1 303 Intron XP_016857289.1
XM_017001801.1 303 Intron XP_016857290.1
XM_017001802.1 303 Intron XP_016857291.1
XM_017001803.1 303 Missense Mutation CAT,CGT H49R XP_016857292.1
XM_017001804.1 303 Intron XP_016857293.1
XM_017001805.1 303 Intron XP_016857294.1

View Full Product Details