Product Details

SNP ID
rs146501611
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51022706 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCCCCCGGAGTCACCCTGGGCAC[A/G]GGGAGAGAGAATGTCGGTCAGAGAA
Phenotype
MIM: 602673 MIM: 604434 MIM: 605539
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KLK10 PubMed Links

Gene Details

Gene
KLK10
Gene Name
kallikrein related peptidase 10
There are no transcripts associated with this gene.

Gene
KLK11
Gene Name
kallikrein related peptidase 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136032.2 Intron NP_001129504.1
NM_001167605.1 Intron NP_001161077.1
NM_006853.2 Intron NP_006844.1
NM_144947.1 Intron NP_659196.1
XM_005258439.3 Intron XP_005258496.1
XM_011526369.1 Intron XP_011524671.1
XM_011526370.2 Intron XP_011524672.1
XM_011526371.2 Intron XP_011524673.1
XM_011526372.2 Intron XP_011524674.1
XM_011526373.1 Intron XP_011524675.1
Gene
KLK12
Gene Name
kallikrein related peptidase 12
There are no transcripts associated with this gene.

View Full Product Details