Product Details

SNP ID
rs61729817
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51023193 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACACTTCTGGTGCTCAATGATGG[C/T]GATGTTGGCGCATCGCAAGGTGTGA
Phenotype
MIM: 602673 MIM: 604434 MIM: 605539
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLK10 PubMed Links

Gene Details

Gene
KLK10
Gene Name
kallikrein related peptidase 10
There are no transcripts associated with this gene.

Gene
KLK11
Gene Name
kallikrein related peptidase 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136032.2 628 Missense Mutation NP_001129504.1
NM_001167605.1 628 Missense Mutation NP_001161077.1
NM_006853.2 628 Missense Mutation NP_006844.1
NM_144947.1 628 Missense Mutation NP_659196.1
XM_005258439.3 628 Missense Mutation XP_005258496.1
XM_011526369.1 628 Missense Mutation XP_011524671.1
XM_011526370.2 628 Missense Mutation XP_011524672.1
XM_011526371.2 628 Missense Mutation XP_011524673.1
XM_011526372.2 628 Missense Mutation XP_011524674.1
XM_011526373.1 628 Missense Mutation XP_011524675.1
Gene
KLK12
Gene Name
kallikrein related peptidase 12
There are no transcripts associated with this gene.

View Full Product Details