Product Details

SNP ID
rs60638904
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:50378328 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCAGTGTGGTCCGTGGTGGGGCC[A/C]GGCGCTATGCCTGCCGGGGTGGCGG
Phenotype
MIM: 605631 MIM: 600380 MIM: 174761
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
NAPSA PubMed Links

Gene Details

Gene
NAPSA
Gene Name
napsin A aspartic peptidase
There are no transcripts associated with this gene.

Gene
NR1H2
Gene Name
nuclear receptor subfamily 1 group H member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256647.1 642 Intron NP_001243576.1
NM_007121.5 642 Silent Mutation AGG,CGG R121R NP_009052.3
Gene
POLD1
Gene Name
DAN polymerase delta 1, catalytic subunit
There are no transcripts associated with this gene.

View Full Product Details