Product Details

SNP ID
rs45593434
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:31335917 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTATGTTCCTCCTGCTCCATGGAAG[C/G]CCAAAGGCAGCACAAGAAGACTCTG
Phenotype
MIM: 607633
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
XDH PubMed Links

Gene Details

Gene
XDH
Gene Name
xanthine dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000379.3 4122 UTR 3 NP_000370.2
XM_011533095.2 4122 UTR 3 XP_011531397.1
XM_011533096.2 4122 Intron XP_011531398.1

View Full Product Details