Product Details

SNP ID
rs750118212
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:102398560 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAGGACCTCAGGGTGCTGGCGGG[A/G]GGCCAGGCTGGGCTAGAAGAAGGTC
Phenotype
MIM: 164012 MIM: 602327
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
NFKB2 PubMed Links

Gene Details

Gene
NFKB2
Gene Name
nuclear factor kappa B subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077494.3 Intron NP_001070962.1
NM_001261403.2 Intron NP_001248332.1
NM_001288724.1 Intron NP_001275653.1
NM_001322934.1 Intron NP_001309863.1
NM_001322935.1 Intron NP_001309864.1
NM_002502.5 Intron NP_002493.3
XM_011539830.2 Intron XP_011538132.1
XM_011539831.2 Intron XP_011538133.1
XM_017016278.1 Intron XP_016871767.1
Gene
PSD
Gene Name
pleckstrin and Sec7 domain containing
There are no transcripts associated with this gene.

View Full Product Details