Product Details

SNP ID
rs3814556
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.10:97616395 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGCGGGGGATTCCATGAGAACCATC[A/G]GGGAAAGTCAGCAGGCCTTTGAGGA
Phenotype
MIM: 613597
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HOGA1 PubMed Links

Gene Details

Gene
HOGA1
Gene Name
4-hydroxy-2-oxoglutarate aldolase 1
There are no transcripts associated with this gene.

Gene
MORN4
Gene Name
MORN repeat containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098831.1 614 Intron NP_001092301.1
NM_178832.3 614 Silent Mutation CCC,CCT P103P NP_849154.1
XM_011539250.2 614 Silent Mutation CCC,CCT P161P XP_011537552.1
XM_011539251.2 614 Silent Mutation CCC,CCT P103P XP_011537553.1

View Full Product Details