Product Details

SNP ID
rs11039155
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:47259211 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTTGGTAATGACCAGGGCTCCAG[A/G]AAGAGATGTCCTTGTGGCTGGGGGC
Phenotype
MIM: 602423
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NR1H3 PubMed Links

Gene Details

Gene
NR1H3
Gene Name
nuclear receptor subfamily 1 group H member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130101.2 198 UTR 5 NP_001123573.1
NM_001130102.2 198 Intron NP_001123574.1
NM_001251934.1 198 Intron NP_001238863.1
NM_001251935.1 198 Intron NP_001238864.1
NM_005693.3 198 UTR 5 NP_005684.2
XM_005252705.1 198 UTR 5 XP_005252762.1
XM_005252706.1 198 UTR 5 XP_005252763.1
XM_005252707.4 198 UTR 5 XP_005252764.1
XM_005252709.1 198 UTR 5 XP_005252766.1
XM_005252710.1 198 UTR 5 XP_005252767.1
XM_005252713.3 198 UTR 5 XP_005252770.1
XM_005252715.2 198 UTR 5 XP_005252772.1
XM_005252716.3 198 Intron XP_005252773.1
XM_005252718.3 198 Intron XP_005252775.1
XM_006718112.1 198 UTR 5 XP_006718175.1
XM_006718113.1 198 UTR 5 XP_006718176.1
XM_006718115.1 198 UTR 5 XP_006718178.1
XM_006718116.1 198 Intron XP_006718179.1
XM_011519805.2 198 UTR 5 XP_011518107.1
XM_011519806.1 198 Intron XP_011518108.1
XM_011519807.1 198 Intron XP_011518109.1
XM_011519808.2 198 Intron XP_011518110.1
XM_017017056.1 198 Intron XP_016872545.1
XM_017017057.1 198 Intron XP_016872546.1
XM_017017058.1 198 Intron XP_016872547.1

View Full Product Details