Product Details

SNP ID
rs34392570
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:35663285 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAAGTTCCTCAGTCACCATCTGGC[C/T]GAATACGTCCACGGCTCCCAGGCCT
Phenotype
MIM: 612298
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRIM44 PubMed Links

Gene Details

Gene
TRIM44
Gene Name
tripartite motif containing 44
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017583.5 534 Silent Mutation GCC,GCT A58A NP_060053.2
XM_006718254.1 534 Silent Mutation GCC,GCT A58A XP_006718317.1

View Full Product Details