Product Details
- SNP ID
-
rs78256349
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
13
- Location
-
Chr.1:10461586 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCTCCGGAGAGAATGCAAGCTCTG[C/T]GTCTGCTGCAGGCGCAGGGCGAGCT
- Phenotype
-
MIM: 602784
MIM: 601882
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
APITD1-CORT
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75956311] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- APITD1-CORT
- Gene Name
- APITD1-CORT readthrough
There are no transcripts associated with this gene.
- Gene
- CORT
- Gene Name
- cortistatin
There are no transcripts associated with this gene.
- Gene
- DFFA
- Gene Name
- DNA fragmentation factor subunit alpha
View Full Product Details