Product Details

SNP ID
rs34558933
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:106806737 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTGCACCGCCTGGAAGCCGGGAA[C/G]AACCACCTTGCCCTTGCGATGCAAA
Phenotype
MIM: 608472
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ST6GAL2 PubMed Links

Gene Details

Gene
ST6GAL2
Gene Name
ST6 beta-galactoside alpha-2,6-sialyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142351.1 1650 Missense Mutation CTT,GTT L511V NP_001135823.1
NM_001142352.1 1650 Intron NP_001135824.1
NM_001322362.1 1650 Missense Mutation CTT,GTT L511V NP_001309291.1
NM_032528.2 1650 Missense Mutation CTT,GTT L511V NP_115917.1
XM_006712802.1 1650 Missense Mutation CTT,GTT L511V XP_006712865.1
XM_011511999.2 1650 Missense Mutation CTT,GTT L511V XP_011510301.1
XM_011512000.2 1650 Intron XP_011510302.1
XM_011512001.2 1650 Intron XP_011510303.1
XM_017005110.1 1650 Intron XP_016860599.1

View Full Product Details