Product Details

SNP ID
rs111877081
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75012951 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTAGGAACCCCAGGCCCATTGGCC[A/G]GATGTGGAGCTACGTCGGGGGGCTA
Phenotype
MIM: 603000
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDR2L PubMed Links

Gene Details

Gene
CDR2L
Gene Name
cerebellar degeneration related protein 2 like
There are no transcripts associated with this gene.

Gene
MRPL58
Gene Name
mitochondrial ribosomal protein L58
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303265.1 Intron NP_001290194.1
NM_001545.2 Intron NP_001536.1
XM_017024574.1 Intron XP_016880063.1

View Full Product Details