Product Details

SNP ID
rs62638692
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:82658013 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTGCATCATCCTGGCATTCAGGCC[A/G]TTGGCCATCGAGAAGGACTTGAGGT
Phenotype
MIM: 609226
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAB40B PubMed Links

Gene Details

Gene
RAB40B
Gene Name
RAB40B, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006822.2 925 Silent Mutation AAC,AAT N229N NP_006813.1
XM_005256334.4 925 Silent Mutation AAC,AAT N169N XP_005256391.1
XM_005256335.3 925 Intron XP_005256392.1
XM_006722271.3 925 Silent Mutation AAC,AAT N189N XP_006722334.1
XM_011523528.2 925 Silent Mutation AAC,AAT N212N XP_011521830.1
XM_017024042.1 925 Intron XP_016879531.1
XM_017024043.1 925 Silent Mutation AAC,AAT N169N XP_016879532.1
Gene
WDR45B
Gene Name
WD repeat domain 45B
There are no transcripts associated with this gene.

View Full Product Details