Product Details

SNP ID
rs115048121
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:89316836 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATATCCAGCGCTTCACCTGAAAGA[C/T]AGTGCAAATTGGTTAGGATGCCACC
Phenotype
MIM: 611360 MIM: 174763
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FANCI PubMed Links

Gene Details

Gene
FANCI
Gene Name
Fanconi anemia complementation group I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001113378.1 4474 UTR 3 NP_001106849.1
NM_018193.2 4474 UTR 3 NP_060663.2
XM_011521756.2 4474 UTR 3 XP_011520058.1
XM_011521757.2 4474 UTR 3 XP_011520059.1
XM_011521764.2 4474 UTR 3 XP_011520066.1
XM_011521767.2 4474 UTR 3 XP_011520069.1
Gene
MIR6766
Gene Name
microRNA 6766
There are no transcripts associated with this gene.

Gene
POLG
Gene Name
DNA polymerase gamma, catalytic subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001126131.1 4474 Intron NP_001119603.1
NM_002693.2 4474 Intron NP_002684.1

View Full Product Details