Product Details

SNP ID
rs8045571
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:70253671 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAGGCCACATGTCAGCCACCAGAG[G/A]GCTGATGAGCCCTAGGGGAGGGGAC
Phenotype
MIM: 601065 MIM: 606490
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
AARS PubMed Links

Gene Details

Gene
AARS
Gene Name
alanyl-tRNA synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001605.2 Intron NP_001596.2
Gene
EXOSC6
Gene Name
exosome component 6
There are no transcripts associated with this gene.

Gene
LOC107984138
Gene Name
serine/threonine-protein kinase SMG1-like
There are no transcripts associated with this gene.

View Full Product Details