Product Details
- SNP ID
-
rs13339140
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:67180020 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCCTTGGGTGGTCAGGGCCAGGATA[C/T]GGTGGGGAGGCTGGACTGACCTGAT
- Phenotype
-
MIM: 614117
MIM: 605235
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EXOC3L1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115387781] are located under a probe and SNP(s) [rs61733768] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EXOC3L1
- Gene Name
- exocyst complex component 3 like 1
There are no transcripts associated with this gene.
- Gene
- KIAA0895L
- Gene Name
- KIAA0895 like
- Gene
- NOL3
- Gene Name
- nucleolar protein 3
There are no transcripts associated with this gene.
View Full Product Details