Product Details

SNP ID
rs34392264
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:40729507 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAAAGGCCACAAGTCACCAGCTC[C/T]ATGAAGTGGCGAATTGGTCCTTGTT
Phenotype
MIM: 611992
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR320D1 PubMed Links

Gene Details

Gene
MIR320D1
Gene Name
microRNA 320d-1
There are no transcripts associated with this gene.

Gene
MRPS31
Gene Name
mitochondrial ribosomal protein S31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005830.3 1128 Missense Mutation ATA,ATG I351M NP_005821.2

View Full Product Details