Product Details

SNP ID
hCV25615051
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:145826795 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGAGCTATCTTAGGCCATCTCAT[C/A]TGCCTTTTTCCTCCTGTTATACAGA
Phenotype
Polymorphism
C/A, Transversion substitution
Allele Nomenclature
Literature Links
POLR3C PubMed Links

Gene Details

Gene
POLR3C
Gene Name
RNA polymerase III subunit C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303456.1 Intron NP_001290385.1
NM_006468.7 Intron NP_006459.3
Gene
RNF115
Gene Name
ring finger protein 115
There are no transcripts associated with this gene.

View Full Product Details