Product Details

SNP ID
rs34347676
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:76954117 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGCGCAGCCTTCTTCATCAGCGA[A/G]CCCGAGGAGTTCATTACCATCCACT
Phenotype
MIM: 122559 MIM: 610103
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CRHBP PubMed Links

Gene Details

Gene
CRHBP
Gene Name
corticotropin releasing hormone binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001882.3 544 Silent Mutation GAA,GAG E88E NP_001873.2
Gene
S100Z
Gene Name
S100 calcium binding protein Z
There are no transcripts associated with this gene.

View Full Product Details