Product Details

SNP ID
rs79541226
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:170256427 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGAGGAAATCAGATGCTTTTAGG[A/T]AACAATAAAACCTTTGTCGAAAGCT
Phenotype
MIM: 601603
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
C5orf58 PubMed Links

Gene Details

Gene
C5orf58
Gene Name
chromosome 5 open reading frame 58
There are no transcripts associated with this gene.

Gene
LCP2
Gene Name
lymphocyte cytosolic protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005565.3 Intron NP_005556.1

View Full Product Details