Product Details
- SNP ID
-
rs5745297
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:10680885 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTCCCCATAAACAAGGTTTGGGCTG[A/G]AGCTGTTTCTAGTTTTAAATATGGA
- Phenotype
-
MIM: 600954
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DAP
PubMed Links
Gene Details
- Gene
- DAP
- Gene Name
- death associated protein
View Full Product Details