Product Details

SNP ID
rs4988492
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:37254295 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACATGCTGTCTACCTGACGACCAA[A/G]CCGTGCCCTTGCTCCTCGCTCTTGG
Phenotype
MIM: 139190
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GHRH PubMed Links

Gene Details

Gene
GHRH
Gene Name
growth hormone releasing hormone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184731.2 242 Missense Mutation CTT,TTT L75F NP_001171660.1
NM_021081.5 242 Missense Mutation CTT,TTT L75F NP_066567.1
XM_011528784.2 242 Missense Mutation CTT,TTT L75F XP_011527086.1
XM_011528788.2 242 Missense Mutation CTT,TTT L75F XP_011527090.1

View Full Product Details