Product Details

SNP ID
hCV25619617
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:27711145 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCACCCGCGTTGCCCGGCTTCTA[A/G]CCGCCTGTTTCTGCCCTGGGTGCTC
Phenotype
MIM: 611995
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPS35 PubMed Links

Gene Details

Gene
MRPS35
Gene Name
mitochondrial ribosomal protein S35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190864.1 Intron NP_001177793.1
NM_021821.3 Intron NP_068593.2

View Full Product Details