Product Details

SNP ID
rs41302643
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:11763612 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTTTCTGCACTTTATGTTTTCCT[A/G]TGTAATCTTGACAGTAGCTACAAAT
Phenotype
MIM: 300609
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MSL3 PubMed Links

Gene Details

Gene
MSL3
Gene Name
male-specific lethal 3 homolog (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193270.2 Intron NP_001180199.1
NM_001282174.1 Intron NP_001269103.1
NM_006800.3 Intron NP_006791.2
NM_078628.1 Intron NP_523352.1
NM_078629.3 Intron NP_523353.2
XM_005274440.2 Intron XP_005274497.1
XM_011545442.2 Intron XP_011543744.1

View Full Product Details