Product Details

SNP ID
rs3736009
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:142735284 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTGGGCAGCGGCCCAGAGGTGGGA[A/C]GGCCGTAGCTTAGCTGGGTGTCCCC
Phenotype
MIM: 606119
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC100288181 PubMed Links

Gene Details

Gene
LOC100288181
Gene Name
uncharacterized LOC100288181
There are no transcripts associated with this gene.

Gene
SLURP1
Gene Name
secreted LY6/PLAUR domain containing 1
There are no transcripts associated with this gene.

Gene
THEM6
Gene Name
thioesterase superfamily member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016647.2 Intron NP_057731.1
XM_005250955.2 Intron XP_005251012.1

View Full Product Details