Product Details

SNP ID
rs36054034
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:151103597 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATGGCTTGGTGGAGAGACTTGTC[A/G]TATTTCTCAATGAATTCCCTCCGGA
Phenotype
MIM: 114070
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANXA6 PubMed Links

Gene Details

Gene
ANXA6
Gene Name
annexin A6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001155.4 2145 Silent Mutation TAC,TAT Y645Y NP_001146.2
NM_001193544.1 2145 Silent Mutation TAC,TAT Y613Y NP_001180473.1
XM_005268432.4 2145 Silent Mutation TAC,TAT Y639Y XP_005268489.1

View Full Product Details