Product Details

SNP ID
rs11880898
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44118768 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTTTTCCTCATGGCAGCCAATGTA[A/T]TCCTTAGGAAAGTCAGACTGCCATT
Phenotype
MIM: 194555
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC100379224 PubMed Links

Gene Details

Gene
LOC100379224
Gene Name
uncharacterized LOC100379224
There are no transcripts associated with this gene.

Gene
ZNF224
Gene Name
zinc finger protein 224
There are no transcripts associated with this gene.

Gene
ZNF225
Gene Name
zinc finger protein 225
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321685.1 Intron NP_001308614.1
NM_013362.3 Intron NP_037494.2
XM_005259223.3 Intron XP_005259280.1
XM_011527285.2 Intron XP_011525587.1
XM_011527286.2 Intron XP_011525588.1

View Full Product Details