Product Details

SNP ID
rs62640920
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:13849533 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAAAATCAGCCCAGGAACAATGAG[A/G]CCAATCACAAATAAGTCCCAGTGTG
Phenotype
MIM: 614289
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SEL1L2 PubMed Links

Gene Details

Gene
SEL1L2
Gene Name
SEL1L2 ERAD E3 ligase adaptor subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271539.1 1860 Silent Mutation GGC,GGT G560G NP_001258468.1
XM_006723646.2 1860 Silent Mutation GGC,GGT G674G XP_006723709.1
XM_006723647.2 1860 Silent Mutation GGC,GGT G673G XP_006723710.1
XM_006723648.2 1860 Silent Mutation GGC,GGT G655G XP_006723711.1
XM_006723650.2 1860 UTR 3 XP_006723713.1
XM_006723651.2 1860 UTR 3 XP_006723714.1
XM_006723652.2 1860 Silent Mutation GGC,GGT G561G XP_006723715.1
XM_006723654.3 1860 Silent Mutation GGC,GGT G338G XP_006723717.1
XM_006723655.2 1860 Intron XP_006723718.1
XM_006723656.2 1860 Intron XP_006723719.1
XM_011529374.1 1860 Silent Mutation GGC,GGT G654G XP_011527676.1
XM_011529375.1 1860 Silent Mutation GGC,GGT G637G XP_011527677.1
XM_011529376.1 1860 Silent Mutation GGC,GGT G618G XP_011527678.1
XM_011529377.1 1860 Silent Mutation GGC,GGT G589G XP_011527679.1
XM_011529378.1 1860 Silent Mutation GGC,GGT G562G XP_011527680.1
XM_011529379.1 1860 Silent Mutation GGC,GGT G535G XP_011527681.1
XM_017028084.1 1860 Silent Mutation GGC,GGT G323G XP_016883573.1

View Full Product Details