Product Details
- SNP ID
-
hCV25625045
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:17581928 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGTGTTGGATTGTATGATTTTATT[C/T]CACTATTAAGCAGAACATTTAAGAT
- Phenotype
-
MIM: 170250
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LAP3
PubMed Links
Gene Details
- Gene
- LAP3
- Gene Name
- leucine aminopeptidase 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015907.2 |
|
Intron |
|
|
NP_056991.2 |
View Full Product Details