Product Details

SNP ID
rs79829943
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:42529589 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCTTATACTTGCTGAGAGTCATT[C/T]TGGTTACTTACTTTTGTGAAAAGTA
Phenotype
MIM: 602534
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LRRC57 PubMed Links

Gene Details

Gene
LRRC57
Gene Name
leucine rich repeat containing 57
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153260.2 Intron NP_694992.2
XM_011521423.2 Intron XP_011519725.1
XM_011521424.2 Intron XP_011519726.1
Gene
SNAP23
Gene Name
synaptosome associated protein 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003825.3 Intron NP_003816.2
NM_130798.2 Intron NP_570710.1
XM_006720725.2 Intron XP_006720788.1
XM_017022692.1 Intron XP_016878181.1
XM_017022693.1 Intron XP_016878182.1
XM_017022694.1 Intron XP_016878183.1

View Full Product Details