Product Details

SNP ID
rs199523375
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:113410711 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAGGTGGGAAGCAGGCTGCTGTG[C/T]GGGCAGGCAGCAGAGTCAGCAGTGG
Phenotype
MIM: 126450
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DRD2 PubMed Links

Gene Details

Gene
DRD2
Gene Name
dopamine receptor D2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000795.3 1565 UTR 3 NP_000786.1
NM_016574.3 1565 UTR 3 NP_057658.2
XM_017017296.1 1565 UTR 3 XP_016872785.1

View Full Product Details