Product Details

SNP ID
rs17885542
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:24795839 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGAGCGGGGAGGAAGGCAAGGAG[A/G]GGAGGAAGAAACCGTTCCTCCTGTG
Phenotype
MIM: 185490
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SOD3 PubMed Links

Gene Details

Gene
SOD3
Gene Name
superoxide dismutase 3, extracellular
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003102.2 Intron NP_003093.2

View Full Product Details