Product Details

SNP ID
rs61251388
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:41249585 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATCTCATCGTGAAGACCAAGGCT[A/G]CTGGGGCAGACCCAGGCCTCGGGGC
Phenotype
MIM: 616236 MIM: 602362
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHADL PubMed Links

Gene Details

Gene
CHADL
Gene Name
chondroadherin like
There are no transcripts associated with this gene.

Gene
MIR6889
Gene Name
microRNA 6889
There are no transcripts associated with this gene.

Gene
RANGAP1
Gene Name
Ran GTPase activating protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278651.1 Intron NP_001265580.1
NM_001317930.1 Intron NP_001304859.1
NM_002883.3 Intron NP_002874.1
XM_005261695.1 Intron XP_005261752.1
XM_005261696.1 Intron XP_005261753.1
XM_006724289.3 Intron XP_006724352.1
XM_011530293.1 Intron XP_011528595.1
XM_011530294.2 Intron XP_011528596.1
XM_011530295.2 Intron XP_011528597.1
XM_011530297.1 Intron XP_011528599.1
XM_017028893.1 Intron XP_016884382.1
XM_017028894.1 Intron XP_016884383.1
XM_017028895.1 Intron XP_016884384.1
XM_017028896.1 Intron XP_016884385.1
XM_017028897.1 Intron XP_016884386.1
XM_017028898.1 Intron XP_016884387.1
XM_017028899.1 Intron XP_016884388.1

View Full Product Details