Product Details

SNP ID
rs142383251
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:56221520 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCAAATGTCATCTGATAACATTGA[C/T]GAAAAATATCATTCACTTCTTCTTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZSCAN5A PubMed Links

Gene Details

Gene
ZSCAN5A
Gene Name
zinc finger and SCAN domain containing 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001322061.1 1895 UTR 3 NP_001308990.1
NM_001322062.1 1895 Intron NP_001308991.1
NM_001322064.1 1895 UTR 3 NP_001308993.1
NM_001322065.1 1895 UTR 3 NP_001308994.1
NM_001322066.1 1895 UTR 3 NP_001308995.1
NM_001322067.1 1895 UTR 3 NP_001308996.1
NM_001322068.1 1895 UTR 3 NP_001308997.1
NM_001322069.1 1895 UTR 3 NP_001308998.1
NM_001322070.1 1895 UTR 3 NP_001308999.1
NM_001322072.1 1895 UTR 3 NP_001309001.1
NM_001322073.1 1895 UTR 3 NP_001309002.1
NM_001322074.1 1895 UTR 3 NP_001309003.1
NM_001322075.1 1895 UTR 3 NP_001309004.1
NM_001322076.1 1895 UTR 3 NP_001309005.1
NM_001322077.1 1895 UTR 3 NP_001309006.1
NM_001322078.1 1895 UTR 3 NP_001309007.1
NM_024303.2 1895 Intron NP_077279.1
XM_005259254.1 1895 Intron XP_005259311.1
XM_006723377.3 1895 UTR 3 XP_006723440.1
XM_011527308.1 1895 Intron XP_011525610.1
XM_017027298.1 1895 UTR 3 XP_016882787.1
XM_017027299.1 1895 UTR 3 XP_016882788.1

View Full Product Details