Product Details

SNP ID
rs138207690
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:154402704 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCAGGCCCTGGGGGGGTAGGGGGA[C/T]GCTCAGGCAGCAGGGCACAGCTGAG
Phenotype
MIM: 300081 MIM: 312173 MIM: 300394
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNASE1L1 PubMed Links

Gene Details

Gene
DNASE1L1
Gene Name
deoxyribonuclease I-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009932.2 1543 UTR 3 NP_001009932.1
NM_001009933.2 1543 UTR 3 NP_001009933.1
NM_001009934.2 1543 UTR 3 NP_001009934.1
NM_001303620.1 1543 UTR 3 NP_001290549.1
NM_006730.3 1543 UTR 3 NP_006721.1
XM_005277829.4 1543 UTR 3 XP_005277886.1
XM_011531122.2 1543 UTR 3 XP_011529424.1
XM_017029332.1 1543 UTR 3 XP_016884821.1
Gene
LOC107987332
Gene Name
uncharacterized LOC107987332
There are no transcripts associated with this gene.

Gene
RPL10
Gene Name
ribosomal protein L10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256577.2 1543 Intron NP_001243506.2
NM_001256580.2 1543 Intron NP_001243509.2
NM_001303624.1 1543 Intron NP_001290553.1
NM_001303625.1 1543 Intron NP_001290554.1
NM_001303626.1 1543 Intron NP_001290555.1
NM_006013.4 1543 Intron NP_006004.3
Gene
SNORA70
Gene Name
small nucleolar RNA, H/ACA box 70
There are no transcripts associated with this gene.

Gene
TAZ
Gene Name
tafazzin
There are no transcripts associated with this gene.

View Full Product Details