Product Details

SNP ID
rs41292350
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:51978304 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGAAAGGCCTTTAACTCTCACAC[A/G]TGGGAGCAGCTGGGCACACTGCAGC
Phenotype
MIM: 104620
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABHD14A PubMed Links

Gene Details

Gene
ABHD14A
Gene Name
abhydrolase domain containing 14A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015407.4 437 Silent Mutation ACA,ACG T109T NP_056222.2
Gene
ABHD14A-ACY1
Gene Name
ABHD14A-ACY1 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316331.1 437 Missense Mutation CAT,CGT H61R NP_001303260.1
Gene
ABHD14B
Gene Name
abhydrolase domain containing 14B
There are no transcripts associated with this gene.

Gene
ACY1
Gene Name
aminoacylase 1
There are no transcripts associated with this gene.

View Full Product Details