Product Details

SNP ID
rs60059725
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:39335372 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGATCCATGGGGGACACAAGCCC[A/G]GCCCTCCCTATCTGCCCTCACCACC
Phenotype
MIM: 604104
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GMFG PubMed Links

Gene Details

Gene
GMFG
Gene Name
glia maturation factor gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301008.1 Intron NP_001287937.1
NM_004877.3 Intron NP_004868.1
Gene
SAMD4B
Gene Name
sterile alpha motif domain containing 4B
There are no transcripts associated with this gene.

View Full Product Details