Product Details

SNP ID
rs140676747
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:53006459 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGTGATTGCCTCATCCGGGTCTT[C/T]TGCGTTCTCTTTCCCTCTCCCAACA
Phenotype
MIM: 603928
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EIF4B PubMed Links

Gene Details

Gene
EIF4B
Gene Name
eukaryotic translation initiation factor 4B
There are no transcripts associated with this gene.

View Full Product Details