Product Details

SNP ID
rs61730653
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:112822146 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAATCAGGTGTGATGATTAGTAA[C/T]AAACCATTTAACACTAACAGTTCAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BMT2 PubMed Links

Gene Details

Gene
BMT2
Gene Name
base methyltransferase of 25S rRNA 2 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152556.2 943 Silent Mutation TTA,TTG L272L NP_689769.2

View Full Product Details