Product Details

SNP ID
rs61730659
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:36135780 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGGTGATGCCCTTCATGCAGAC[A/G]GATCTGCAGAAGATCATGGGGATGG
Phenotype
MIM: 602899
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAPK13 PubMed Links

Gene Details

Gene
MAPK13
Gene Name
mitogen-activated protein kinase 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002754.4 435 Silent Mutation ACA,ACG T112T NP_002745.1

View Full Product Details