Product Details
- SNP ID
-
rs146427285
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:157734504 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TACTCTCTGTGGTACCCACAGGTCT[C/T]ATCATGGTGTTGAACTAATTCCGTG
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LSM11
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115847385] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LSM11
- Gene Name
- LSM11, U7 small nuclear RNA associated
There are no transcripts associated with this gene.
- Gene
- THG1L
- Gene Name
- tRNA-histidine guanylyltransferase 1 like
View Full Product Details