Product Details

SNP ID
rs17398660
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:125032195 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTGGTAATGTGAGGTTTAAGTTAG[A/G]TTTTGTGACAAGAAGTCCCACGTAG
Phenotype
MIM: 610657 MIM: 602019
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIAA0196 PubMed Links

Gene Details

Gene
KIAA0196
Gene Name
KIAA0196
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014846.3 Intron NP_055661.3
XM_005251120.3 Intron XP_005251177.1
XM_011517409.1 Intron XP_011515711.1
XM_017014113.1 Intron XP_016869602.1
Gene
SQLE
Gene Name
squalene epoxidase
There are no transcripts associated with this gene.

View Full Product Details