Product Details

SNP ID
rs186785842
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:216109380 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGCTTGTCCACCGGAAGCGAGTT[A/G]CGACACGGCAGGTTCCCGCCCGGAA
Phenotype
MIM: 194364
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM169 PubMed Links

Gene Details

Gene
TMEM169
Gene Name
transmembrane protein 169
There are no transcripts associated with this gene.

Gene
XRCC5
Gene Name
X-ray repair cross complementing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021141.3 84 UTR 5 NP_066964.1

View Full Product Details