Product Details

SNP ID
rs36081973
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:44885365 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATTGACTTCTTGAATCAGGACAA[C/T]GCCGTTTCTCACCACACATGGGAGT
Phenotype
MIM: 600585
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TGM4 PubMed Links

Gene Details

Gene
TGM4
Gene Name
transglutaminase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003241.3 133 Silent Mutation AAC,AAT N20N NP_003232.2
XM_011534042.1 133 Silent Mutation AAC,AAT N65N XP_011532344.1

View Full Product Details