Product Details
- SNP ID
-
rs147626572
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:133304659 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCCTCGGTTGACAATTGAGAGAAC[C/T]CTCTCTGAAGGTCACCCAGGCATCT
- Phenotype
-
MIM: 300168
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GPC4
PubMed Links
Gene Details
- Gene
- GPC4
- Gene Name
- glypican 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001448.2 |
|
Intron |
|
|
NP_001439.2 |
View Full Product Details