Product Details
- SNP ID
-
rs61732846
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:148811688 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGGATCTTTGATAAAAATCCCCCA[C/G]CCTGCCACGTCAGATGGTGCCAGCA
- Phenotype
-
MIM: 601573
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
EZH2
PubMed Links
Gene Details
- Gene
- EZH2
- Gene Name
- enhancer of zeste 2 polycomb repressive complex 2 subunit
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001203247.1 |
4403 |
Silent Mutation |
GGC,GGG |
G623G |
NP_001190176.1 |
NM_001203248.1 |
4403 |
Silent Mutation |
GGC,GGG |
G614G |
NP_001190177.1 |
NM_001203249.1 |
4403 |
Silent Mutation |
GGC,GGG |
G572G |
NP_001190178.1 |
NM_004456.4 |
4403 |
Silent Mutation |
GGC,GGG |
G628G |
NP_004447.2 |
NM_152998.2 |
4403 |
Silent Mutation |
GGC,GGG |
G584G |
NP_694543.1 |
XM_005249962.4 |
4403 |
Silent Mutation |
GGC,GGG |
G631G |
XP_005250019.1 |
XM_005249963.4 |
4403 |
Silent Mutation |
GGC,GGG |
G622G |
XP_005250020.1 |
XM_005249964.4 |
4403 |
Silent Mutation |
GGC,GGG |
G580G |
XP_005250021.1 |
XM_011515883.2 |
4403 |
Silent Mutation |
GGC,GGG |
G636G |
XP_011514185.1 |
XM_011515884.2 |
4403 |
Silent Mutation |
GGC,GGG |
G628G |
XP_011514186.1 |
XM_011515885.2 |
4403 |
Silent Mutation |
GGC,GGG |
G627G |
XP_011514187.1 |
XM_011515886.2 |
4403 |
Silent Mutation |
GGC,GGG |
G620G |
XP_011514188.1 |
XM_011515887.2 |
4403 |
Silent Mutation |
GGC,GGG |
G619G |
XP_011514189.1 |
XM_011515888.2 |
4403 |
Silent Mutation |
GGC,GGG |
G619G |
XP_011514190.1 |
XM_011515889.2 |
4403 |
Silent Mutation |
GGC,GGG |
G606G |
XP_011514191.1 |
XM_011515890.2 |
4403 |
Silent Mutation |
GGC,GGG |
G597G |
XP_011514192.1 |
XM_011515891.2 |
4403 |
Silent Mutation |
GGC,GGG |
G595G |
XP_011514193.1 |
XM_011515892.2 |
4403 |
Silent Mutation |
GGC,GGG |
G594G |
XP_011514194.1 |
XM_011515893.2 |
4403 |
Silent Mutation |
GGC,GGG |
G592G |
XP_011514195.1 |
XM_011515894.2 |
4403 |
Silent Mutation |
GGC,GGG |
G589G |
XP_011514196.1 |
XM_011515895.2 |
4403 |
Silent Mutation |
GGC,GGG |
G588G |
XP_011514197.1 |
XM_011515896.2 |
4403 |
Silent Mutation |
GGC,GGG |
G550G |
XP_011514198.1 |
XM_011515897.2 |
4403 |
Silent Mutation |
GGC,GGG |
G519G |
XP_011514199.1 |
XM_011515898.2 |
4403 |
Silent Mutation |
GGC,GGG |
G519G |
XP_011514200.1 |
XM_011515899.2 |
4403 |
Intron |
|
|
XP_011514201.1 |
XM_011515901.2 |
4403 |
Intron |
|
|
XP_011514203.1 |
XM_017011817.1 |
4403 |
Silent Mutation |
GGC,GGG |
G636G |
XP_016867306.1 |
XM_017011818.1 |
4403 |
Silent Mutation |
GGC,GGG |
G615G |
XP_016867307.1 |
XM_017011819.1 |
4403 |
Silent Mutation |
GGC,GGG |
G589G |
XP_016867308.1 |
XM_017011820.1 |
4403 |
Silent Mutation |
GGC,GGG |
G580G |
XP_016867309.1 |
XM_017011821.1 |
4403 |
Silent Mutation |
GGC,GGG |
G514G |
XP_016867310.1 |
View Full Product Details